missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Piperidinecarboxylate-2,2,3,3,4,5,5,6,6-d9, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Ethyl Isonipecotate-2,2,3,3,4,5,5,6,6-D9 |
| CAS | 1219803-75-4 |
| Molecular Formula | C8 D9 H6 N O2 |
| Quantity | 0.05 g |
| Synonym | 4-(Ethoxycarbonyl)piperidine-2,2,3,3,4,5,5,6,6-d9, 4-Carboethoxypiperidine-d9, Ethyl 4-piperidinecarboxylate-d9, Ethyl 4-Piperidinecarboxylate-2,2,3,3,4,5,5,6,6-d9, Ethyl isonipecotate-d9, Piperidin-4-carboxylic-2,2,3,3,4,5,5,6,6-d9 acid ethyl ester, 4-Piperidinecarboxylic-2,2,3,3,4,5,5,6,6-d9 acid ethyl ester, Isonipecotic-2,2,3,3,4,5,5,6,6-d9 acid ethyl ester |
| InChI Formula | InChI=1 S/C8H15NO2/c1-2-11-8(10)7-3-5-9-6-4-7/h7,9 H,2-6H2,1H3/i3D2,4D2,5D2,6D2,7 D |
| SMILES | [2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])C([2 H])(C(=O)OCC)C1([2 H])[2 H] |
| IUPAC Name | ethyl 2,2,3,3,4,5,5,6,6-nonadeuteriopiperidine-4-carboxylate |
| Molecular Weight (g/mol) | 166.2657 |
| Formula Weight | 166.1668 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction