Learn More
(±)-Glyceryl-1,1,2,3,3-d5 1-Monooleate, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | 1-Oleoyl Glycerol-d5 |
| CAS | 565183-24-6 |
| Molecular Formula | C21 D5 H35 O4 |
| Quantity | 0.05 g |
| Synonym | 9-Octadecenoic acid (9 Z)-, 2,3-dihydroxypropyl-1,1,2,3,3-d5 ester (9 CI, ACI), 2,3-Dihydroxypropyl-1,1,2,3,3-d5 (9 Z)-9-octadecenoate (ACI), (±)-Glyceryl-1,1,2,3,3-d5 1-Monooleate, (9 Z)-9-Octadecenoic acid 2,3-dihydroxypropyl-1,1,2,3,3-d5 ester, α-Glyceryl-d5 monooloeate, α-Monoolein-d5, Glyceryl 1-Monooleate-D5 (glyceryl-D5) |
| InChI Formula | InChI=1 S/C21H40O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h9-10,20,22-23 H,2-8,11-19H2,1H3/b10-9-/i18D2,19D2,20 D |
| SMILES | [2 H]C([2 H])(O)C([2 H])(O)C([2 H])([2 H])OC(=O)CCCCCCC\C=C/CCCCCCCC |
| IUPAC Name | (1,1,2,3,3-pentadeuterio-2,3-dihydroxypropyl) (Z)-octadec-9-enoate |
| Molecular Weight (g/mol) | 361.5707 |
| Formula Weight | 361.324 g/mol |
| Show More |