missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanoyl-L-carnitine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Palmitoyl-L-carnitine-d3 Hydrochloride |
| CAS | 1334532-26-1 |
| Molecular Formula | C23 D3 H43 N O4 . Cl |
| Quantity | 0.05 g |
| Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexadecyl)oxy]-1-propanaminium-d3 Chloride, L-(3-Carboxy-2-hydroxypropyl)trimethyl-ammonium-d3 Chloride Palmitate, L-Palmitic acid ester with (3-carboxy-2-hydroxypropyl)trimethylammonium-d3 chloride, (-)-(3-Carboxy-2-hydroxypropyl)trimethyl-ammonium-d3 Chloride Palmitate, Hexadecanoyl-L-carnitine-d3 hydrochloride, L-Palmitoylcarnitine-d3 chloride, Hexadecanoyl-L-carnitine-d3 HCl (N-methyl-d3) |
| InChI Formula | InChI=1 S/C23H45NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(27)28-21(19-22(25)26)20-24(2,3)4;/h21H,5-20H2,1-4H3;1 H/t21-;/m1./s1/i2D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCCCCCCCCCC |
| IUPAC Name | [(2 R)-3-carboxy-2-hexadecanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 439.09 |
| Formula Weight | 438.3304 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction