missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Imazalil-d5 Sulfate (allyloxy-d5), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Imazalil-d5 Sulfate |
| CAS | 1398065-92-3 |
| Molecular Formula | C14 D5 H9 Cl2 N2 O . H2 O4 S |
| Quantity | 0.05 g |
| Synonym | 1-[2-(2,4-Dichlorophenyl)-2-(1,1,2,3,3-pentadeuterioprop-2-enoxy)ethyl]imidazole sulfuric acid, Imazalil-d5 Sulfate, Imazalil-D5 (2-propenyl-D5) Sulfate, (±)-Imazalil-d5 Sulfate (allyloxy-d5), Enilconazole-D5 (allyloxy-D5) Sulfate, 1-[2-(2,4-Dichlorophenyl)-2-(2-propenyloxy-1,1,2,3,3-d5)ethyl]-1 H-imidazole sulfate (1:1), 1-[2-(2,4-Dichlorophenyl)-2-(2-propenyloxy-1,1,2,3,3-d5)ethyl]-1 H-imidazole sulfate (1:1), 1-[2-(2,4-Dichlorophenyl)-2-(2-propenyloxy-1,1,2,3,3-d5)ethyl]-1 H-imidazole sulfate (1:1) |
| InChI Formula | InChI=1 S/C14H14Cl2N2O.H2O4S/c1-2-7-19-14(9-18-6-5-17-10-18)12-4-3-11(15)8-13(12)16;1-5(2,3)4/h2-6,8,10,14 H,1,7,9H2;(H2,1,2,3,4)/i1D2,2 D,7D2; |
| SMILES | [2 H]C(=C([2 H])C([2 H])([2 H])OC(Cn1ccnc1)c2ccc(Cl)cc2Cl)[2 H].OS(=O)(=O)O |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-(1,1,2,3,3-pentadeuterioprop-2-enoxy)ethyl]imidazole;sulfuric acid |
| Molecular Weight (g/mol) | 400.289 |
| Formula Weight | 399.047 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu