missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Citrulline-2,3,3,4,4,5,5-d7, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | L-Citrulline-d7 |
| CAS | 2483831-24-7 |
| Molecular Formula | C6 D7 H6 N3 O3 |
| Quantity | 0.05 g |
| Synonym | (2 S)-2-Amino-5-(carbamoylamino)-2,3,3,4,4,5,5-heptadeuteriopentanoic acid, N5-(Aminocarbonyl)-L-ornithine-2,3,3,4,4,5,5-d7, L-Citrulline-2,3,3,4,4,5,5-D7, L-Citrulline-2,3,3,4,4,5,5-d7, L-Citrulline-d7 |
| InChI Formula | InChI=1 S/C6H13N3O3/c7-4(5(10)11)2-1-3-9-6(8)12/h4H,1-3,7H2,(H,10,11)(H3,8,9,12)/t4-/m0/s1/i1D2,2D2,3D2,4 D |
| SMILES | [2 H]C([2 H])(NC(=O)N)C([2 H])([2 H])C([2 H])([2 H])[C@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-5-(carbamoylamino)-2,3,3,4,4,5,5-heptadeuteriopentanoic acid |
| Molecular Weight (g/mol) | 182.2289 |
| Formula Weight | 182.1396 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction