missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mono-Ethyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Monoethyl Phthalate-d4 |
| CAS | 1219806-03-7 |
| Molecular Formula | C10 2H4 H6 O4 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C10H10O4/c1-2-14-10(13)8-6-4-3-5-7(8)9(11)12/h3-6 H,2H2,1H3,(H,11,12)/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCC)c(C(=O)O)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-ethoxycarbonylbenzoic acid |
| Molecular Weight (g/mol) | 198.21 |
| Formula Weight | 198.083 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction