missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-2-Hydroxyethylpiperazine-N'-ethanesulfonic Acid-d18, CDN
Click to view available options
Quantity:
0.05 g
Unit Size:
0.05g
Specifications
Specifications
| Chemical Name or Material | Hepes (D18) |
| CAS | 203805-87-2 |
| Molecular Formula | C8 2H18 N2 O4 S |
| Quantity | 0.05 g |
| Synonym | 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid |
| InChI Formula | InChI=1 S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,11 D/hD |
| SMILES | [2 H]OC([2 H])([2 H])C([2 H])([2 H])N1C([2 H])([2 H])C([2 H])([2 H])N(C([2 H])([2 H])C([2 H])([2 H])S(=O)(=O)O[2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | deuterio 1,1,2,2-tetradeuterio-2-[2,2,3,3,5,5,6,6-octadeuterio-4-(1,1,2,2-tetradeuterio-2-deuteriooxyethyl)piperazin-1-yl]ethanesulfonate |
| Molecular Weight (g/mol) | 256.415 |
| Formula Weight | 256.212 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction