Learn More
N-Nitrosodi-n-propyl-d14-amine, CDN
Specifications
Specifications
| Chemical Name or Material | N-Nitroso-di-n-propylamine D14 |
| CAS | 93951-96-3 |
| Molecular Formula | C6 2H14 N2 O |
| Quantity | 0.05 g |
| Synonym | 1-Propan-1,1,2,2,3,3,3-d7-amine, N-nitroso-N-(propyl-d7)- (9 CI), N-Nitroso-di-n-propylamine D14, N-Nitrosodi-N-propylamine-d14 |
| InChI Formula | InChI=1 S/C6H14N2O/c1-3-5-8(7-9)6-4-2/h3-6H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N(N=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N-bis(1,1,2,2,3,3,3-heptadeuteriopropyl)nitrous amide |
| Molecular Weight (g/mol) | 144.27 |
| Formula Weight | 144.1985 g/mol |
| Show More |