Learn More
Parathion-d10 (diethyl-d10), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Parathion-ethyl D10 (diethyl D10) |
| CAS | 350820-04-1 |
| Molecular Formula | C10 2H10 H4 N O5 P S |
| Quantity | 0.05 g |
| Synonym | Phosphorothioic acid, O,O-di(ethyl-1,1,2,2,2-d5) O-(4-nitrophenyl) ester, Phosphorothioic acid, O,O-di(ethyl-d5) O-(4-nitrophenyl) ester (9 CI), O,O-Di(ethyl-1,1,2,2,2-d5) O-(4-nitrophenyl) phosphorothioate, Parathion-ethyl D10 (diethyl D10) |
| InChI Formula | InChI=1 S/C10H14NO5PS/c1-3-14-17(18,15-4-2)16-10-7-5-9(6-8-10)11(12)13/h5-8 H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])OP(=S)(Oc1ccc(cc1)[N+](=O)[O-])OC([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | (4-nitrophenoxy)-bis(1,1,2,2,2-pentadeuterioethoxy)-sulfanylidene-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 301.32 |
| Formula Weight | 301.0958 g/mol |
| Show More |