Learn More
Tebufenozide-d9 (3,5-dimethylbenzoyl-d9), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Tebufenozide-d9 |
| CAS | 2469006-89-9 |
| Molecular Formula | C22 D9 H19 N2 O2 |
| Quantity | 0.05 g |
| Synonym | N'-(4-Ethylbenzoyl)-N-(2-methylpropan-2-yl)-3,5-bis(trideuteriomethyl)-2,4,6-trideuteriobenzohydrazide, 3,5-Di(methyl-d3)benzoic-2,4,6-d3 acid 1-(1,1-dimethylethyl)-2-(4-ethylbenzoyl)hydrazide, 1-tert-Butyl-1-(3,5-dimethylbenzoyl-d9)-2-(4-ethylbenzoyl)hydrazine, 3,5-Dimethylbenzoic-d9 acid N-tert-butyl-N-(4-ethylbenzoyl)hydrazide, Tebufenozide-d9, Tebufenozide D9 (3,5-Dimethylbenzoyl-D9) |
| InChI Formula | InChI=1 S/C22H28N2O2/c1-7-17-8-10-18(11-9-17)20(25)23-24(22(4,5)6)21(26)19-13-15(2)12-16(3)14-19/h8-14 H,7H2,1-6H3,(H,23,25)/i2D3,3D3,12 D,13 D,14 D |
| SMILES | [2 H]c1c(C(=O)N(NC(=O)c2ccc(CC)cc2)C(C)(C)C)c([2 H])c(c([2 H])c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N-tert-butyl-2,4,6-trideuterio-N'-(4-ethylbenzoyl)-3,5-bis(trideuteriomethyl)benzohydrazide |
| Molecular Weight (g/mol) | 361.525 |
| Formula Weight | 361.272 g/mol |
| Show More |