missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-butyl-d36-ammonium Bromide, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Tetrabutylammonium-D36 Bromide |
| Molecular Formula | C16 D36 N . Br |
| Quantity | 0.05 g |
| Synonym | Tetrabutylammonium-D36 bromide, N,N,N-Tri(butyl-d9)-1-butan-1,1,2,2,3,3,4,4,4-d9-aminium bromide, Tetrabutylammonium-d36 bromide, Tetra-n-butylammonium-d36 bromide |
| InChI Formula | InChI=1 S/C16H36N.BrH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1 H/q+1;/p-1/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[N+](C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])(C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | tetrakis(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)azanium;bromide |
| Molecular Weight (g/mol) | 358.59 |
| Formula Weight | 357.429 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction