Learn More
Tryptamine-alpha,alpha,beta,beta-d4 HCl, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Tryptamine-d4 Hydrochloride |
| CAS | 340257-60-5 |
| Molecular Formula | C10 2H4 H8 N2 . Cl H |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C10H12N2.ClH/c11-6-5-8-7-12-10-4-2-1-3-9(8)10;/h1-4,7,12 H,5-6,11H2;1 H/i5D2,6D2; |
| SMILES | Cl.[2 H]C([2 H])(N)C([2 H])([2 H])c1c[nH]c2ccccc12 |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-(1 H-indol-3-yl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 200.7 |
| Formula Weight | 200.1018 g/mol |
| Percent Purity | 97 atom % D, min 98% Chemical Purity |