missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-d3-2-hexanol-1,1,1,3,3-d5, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-2-hexanol-D8 (1,1,1,2-methyl,3,3-D8) |
| Molecular Formula | C7 D8 H8 O |
| Quantity | 0.1 g |
| Synonym | 2-Methyl-d3-2-hexanol-1,1,1,3,3-d5, 2-(Methyl-d3)-2-hexan-1,1,1,3,3-d5-ol, 2-Methyl-2-hexanol-D8 (1,1,1,2-methyl,3,3-D8), 1,1-Di(methyl-d3)-1-pentanol-2,2-d2, 2-(Methyl-d3)-2-hydroxyhexane-1,1,1,3,3-d5 |
| InChI Formula | InChI=1 S/C7H16O/c1-4-5-6-7(2,3)8/h8H,4-6H2,1-3H3/i2D3,3D3,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C(O)(C([2 H])([2 H])[2 H])C([2 H])([2 H])CCC |
| IUPAC Name | 1,1,1,3,3-pentadeuterio-2-(trideuteriomethyl)hexan-2-ol |
| Molecular Weight (g/mol) | 124.2506 |
| Formula Weight | 124.1703 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction