Learn More
3-Methyl-1-butyl-d11 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 3-Methyl-1-butyl-d11 Alcohol |
| CAS | 170678-50-9 |
| Molecular Formula | C5 D11 H O |
| Quantity | 0.1 g |
| Synonym | 1-Butan-1,1,2,2,3,4,4,4-d8-ol, 3-(methyl-d3)- (9 CI), 3-(Methyl-d3)-1-butan-1,1,2,2,3,4,4,4-d8-ol, 3-Methyl-1-butanol-d11, 3-Methyl-1-butanol-D11, 3-Methyl-1-butyl-d11 Alcohol |
| InChI Formula | InChI=1 S/C5H12O/c1-5(2)3-4-6/h5-6 H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2,5 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,2,3,4,4,4-octadeuterio-3-(trideuteriomethyl)butan-1-ol |
| Molecular Weight (g/mol) | 99.216 |
| Formula Weight | 99.1579 g/mol |
| Show More |