missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylcyclohexanone-2,2,3,3,4,5,5,6,6-d9, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Phenylcyclohexanone-2,2,3,3,4,5,5,6,6-D9 |
| Molecular Formula | C12 D9 H5 O |
| Quantity | 0.1 g |
| Synonym | 4-Phenylcyclohexanone-2,2,3,3,4,5,5,6,6-d9, 4-(Phenyl)cyclohexanone-d9 |
| InChI Formula | InChI=1 S/C12H14O/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-5,11 H,6-9H2/i6D2,7D2,8D2,9D2,11 D |
| SMILES | [2 H]C1([2 H])C(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])(c2ccccc2)C1([2 H])[2 H] |
| IUPAC Name | 2,2,3,3,4,5,5,6,6-nonadeuterio-4-phenylcyclohexan-1-one |
| Molecular Weight (g/mol) | 183.2944 |
| Formula Weight | 183.161 g/mol |
| Percent Purity | 95 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction