missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methyl-d3-mercaptopurine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 6-Methylmercaptopurine-D3 |
| CAS | 33312-93-5 |
| Molecular Formula | C6 D3 H3 N4 S |
| Quantity | 0.1 g |
| Synonym | 9 H-Purine, 6-(methyl-d3-thio)- (ACI), 1 H-Purine, 6-(methyl-d3-thio)- (8 CI, 9 CI), 6-(Methyl-d3-thio)-9 H-purine (ACI), 6-(Methylthio-d3)purine, 6-Methyl-d3-mercaptopurine, 6-Methylmercaptopurine-d3, 6-Methylmercaptopurine-D3 (S-methyl-D3), S-(Methyl-d3)-6-mercaptopurine |
| InChI Formula | InChI=1 S/C6H6N4S/c1-11-6-4-5(8-2-7-4)9-3-10-6/h2-3 H,1H3,(H,7,8,9,10)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])Sc1ncnc2nc[nH]c12 |
| IUPAC Name | 6-(trideuteriomethylsulfanyl)-7 H-purine |
| Molecular Weight (g/mol) | 169.2221 |
| Formula Weight | 169.0501 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction