Learn More
Bis(8-methyl-1-nonyl) Phthalate-3,4,5,6-d4, CDN
Product Code. 30322001
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322001 0.1 g 0.1g
30321642 0.05 g 0.05g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322001 Supplier CDN Supplier No. CDND74520.1G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diisodecyl phthalate-d4
CAS 1346604-79-2
Molecular Formula C28 2H4 H42 O4
Quantity 0.1 g
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-bis(8-methylnonyl) ester, Diisodecyl Phthalate-d4, Phthalic Acid Di-isodecyl Ester (3,4,5,6)-D4
InChI Formula InChI=1 S/C28H46O4/c1-23(2)17-11-7-5-9-15-21-31-27(29)25-19-13-14-20-26(25)28(30)32-22-16-10-6-8-12-18-24(3)4/h13-14,19-20,23-24 H,5-12,15-18,21-22H2,1-4H3/i13D,14 D,19 D,20 D
SMILES [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCCCC(C)C)c(C(=O)OCCCCCCCC(C)C)c1[2 H]
IUPAC Name bis(8-methylnonyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molecular Weight (g/mol) 450.69
Formula Weight 450.3647 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity