missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2-Aminobutyric-2,3,3,4,4,4-d6 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | 2-Aminobutanoic-2,3,3,4,4,4-D6 Acid |
| CAS | 350820-17-6 |
| Molecular Formula | C4H3D6NO2 |
| Quantity | 0.1 g |
| Synonym | Butanoic-2,3,3,4,4,4-d6 acid, 2-amino- (9 CI), 2-Aminobutanoic-2,3,3,4,4,4-d6 acid, DL-2-Aminobutyric acid-d6, DL-Butyrine-d6, DL-Ethylglycine-d6, DL-Homoalanine-d6, DL-α-Aminobutyric acid-d6, 2-Aminobutanoic acid-d6, 2-Aminobutanoic-2,3,3,4,4,4-D6 acid |
| InChI Formula | InChI=1 S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/i1D3,2D2,3 D |
| SMILES | O=C(O)C([2 H])(N)C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | DL-2-Aminobutyric-2,3,3,4,4,4-d6 Acid |
| Molecular Weight (g/mol) | 109.16 |
| Formula Weight | 109.1 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction