Learn More
Hexamethyl-d18-disiloxane, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Hexamethyl-d18-disiloxane |
| CAS | 19108-55-5 |
| Molecular Formula | C6 D18 O Si2 |
| Quantity | 0.1 g |
| Synonym | Disiloxane, 1,1,1,3,3,3-hexa(methyl-d3)- (ACI), 1,1,1,3,3,3-Hexa(methyl-d3)disiloxane (ACI), Disiloxane, hexa(methyl-d3)- (8 CI, 9 CI), Hexamethyl-d18-disiloxane, Hexamethyldisiloxane-d18, Hexamethyldisiloxane-D18 |
| InChI Formula | InChI=1 S/C6H18OSi2/c1-8(2,3)7-9(4,5)6/h1-6H3/i1D3,2D3,3D3,4D3,5D3,6D3 |
| SMILES | [2 H]C([2 H])([2 H])[Si](O[Si](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | tris(trideuteriomethyl)-[tris(trideuteriomethyl)silyloxy]silane |
| Molecular Weight (g/mol) | 180.4884 |
| Formula Weight | 180.2026 g/mol |
| Show More |