Learn More
Hexanoic-5,5-d2 Acid, CDN
Product Code. 30315639
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315639 0.1 g 0.1g
30314191 0.05 g 0.05g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315639 Supplier CDN Supplier No. CDND64200.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Hexanoic-5,5-D2 Acid
CAS 55320-68-8
Molecular Formula C6H10D2O2
Quantity 0.1 g
Synonym Hexanoic-5,5-d2 acid (9 CI), 5,5-Dideuteriohexanoic acid, Hexanoic acid-d2, Hexanoic-5,5-D2 acid, Caproic-delta;-d2 acid
InChI Formula InChI=1 S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i2D2
SMILES CC([2 H])([2 H])CCCC(O)=O
IUPAC Name hexanoic-5,5-d2 acid
Molecular Weight (g/mol) 118.17
Formula Weight 118.1 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity