missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Ibuprofen-d3, Sodium Salt (alpha-methyl-d3), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Ibuprofen-D3 (alpha-methyl-D3) Sodium |
| CAS | 1219805-09-0 |
| Molecular Formula | C13 D3 H14 O2 . Na |
| Quantity | 0.1 g |
| Synonym | 3,3,3-Trideuterio-2-[4-(2-methylpropyl)phenyl]propanoic acid sodium salt, 2-(4-Isobutylphenyl)propanoic-3,3,3-d3 acid sodium salt, α-(Methyl-d3)-4-(2-methylpropyl)benzeneacetic acid sodium salt (1:1), Ibuprofen-D3 (α-methyl-D3) Sodium, Ibuprofen-d3 sodium salt, (±)-Ibuprofen-d3 sodium salt, (±)-Ibuprofen-d3 sodium salt |
| InChI Formula | InChI=1 S/C13H18O2.Na/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15;/h4-7,9-10 H,8H2,1-3H3,(H,14,15);/q;+1/p-1/i3D3; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])C(C(=O)[O-])c1ccc(CC(C)C)cc1 |
| IUPAC Name | sodium;3,3,3-trideuterio-2-[4-(2-methylpropyl)phenyl]propanoate |
| Molecular Weight (g/mol) | 231.2811 |
| Formula Weight | 231.1315 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction