missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium (±)-3-Hydroxybutyrate-3,4,4,4-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | rac 3-Hydroxybutyric Acid-d4 Sodium Salt (BHB-d4) |
| CAS | 1219804-68-8 |
| Molecular Formula | C4 2H4 H3 O3 . Na |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C4H8O3.Na/c1-3(5)2-4(6)7;/h3,5 H,2H2,1H3,(H,6,7);/q;+1/p-1/i1D3,3 D; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])C([2 H])(O)CC(=O)[O-] |
| IUPAC Name | sodium;3,4,4,4-tetradeuterio-3-hydroxybutanoate |
| Molecular Weight (g/mol) | 130.11 |
| Formula Weight | 130.0544 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction