missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10R alpha Antibody (714212), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2742Z
This item is not returnable.
View return policy
Description
IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| IL-10R alpha | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 714212 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
| Mouse myeloma cell line NS0-derived and S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-10 R alpha , His22-Asn235, Accession # Q13651 | |
| 0.1 mL | |
| Cytokine Research | |
| 3587 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction