Learn More
1,1-Dichloroethylene-d2, CDN
Product Code. 30318550
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318550
Brand: CDN CDND22010.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1-Dichloroethene-d2
CAS 22280-73-5
Molecular Formula C2 D2 Cl2
Quantity 0.25 g
Synonym 1,1-Dichloroethylene-d2
InChI Formula InChI=1 S/C2H2Cl2/c1-2(3)4/h1H2/i1D2
SMILES [2 H]C(=C(Cl)Cl)[2 H]
IUPAC Name 1,1-dichloro-2,2-dideuterio-ethylene
Molecular Weight (g/mol) 98.96
Formula Weight 97.966 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity