Learn More
(±)-1,2-Propylene-3,3,3-d3 Oxide, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Propylene Oxide-D3 (methyl-D3) |
| CAS | 2245-32-1 |
| Molecular Formula | C3 D3 H3 O |
| Quantity | 0.25 g |
| Synonym | Oxirane, methyl-d3- (9 CI), Propane-1,1,1-d3, 2,3-epoxy- (7 CI, 8 CI), 1,2-Propylene-3,3,3-d3 oxide, 2-(Methyl-d3)oxirane, Propylene Oxide-D3 (methyl-D3) |
| InChI Formula | InChI=1 S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C1CO1 |
| IUPAC Name | 2-(trideuteriomethyl)oxirane |
| Molecular Weight (g/mol) | 61.0976 |
| Formula Weight | 61.0607 g/mol |
| Show More |