Learn More
2,6-Dimethyl-d6-nitrobenzene, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2,6-Dimethyl-d6-nitrobenzene |
| CAS | 285138-83-2 |
| Molecular Formula | C8 D6 H3 N O2 |
| Quantity | 0.25 g |
| Synonym | Benzene, 1,3-di(methyl-d3)-2-nitro- (9 CI), 1,3-Dimethyl-2-nitrobenzene-D6 (dimethyl-D6), 1,3-Dimethyl-2-nitrobenzene-d6, 2,6-Di(methyl-d3)nitrobenzene, 2-Nitro-1,3-di(methyl-d3)benzene, 2-Nitro-1,3-xylene-d6, 2-Nitro-m-xylene-d6 |
| InChI Formula | InChI=1 S/C8H9NO2/c1-6-4-3-5-7(2)8(6)9(10)11/h3-5 H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])c1cccc(c1[N+](=O)[O-])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-nitro-1,3-bis(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 157.2 |
| Formula Weight | 157.101 g/mol |
| Show More |