missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylpentane-d14, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
missing translation for 'unitSize'
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Methylpentane-D14 |
| CAS | 284487-65-6 |
| Molecular Formula | C6 D14 |
| Quantity | 0.25 g |
| Synonym | Pentane-1,1,1,2,2,3,3,4,5,5,5-d11, 4-(methyl-d3)- (9 CI), 4-(Methyl-d3)pentane-1,1,1,2,2,3,3,4,5,5,5-d11, 2-Methylpentane-d14, 2-Methylpentane-D14 |
| InChI Formula | InChI=1 S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,2,3,3,4,5,5,5-undecadeuterio-4-(trideuteriomethyl)pentane |
| Molecular Weight (g/mol) | 100.2616 |
| Formula Weight | 100.1974 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction