Learn More
Acetamide-2,2,2-d3, CDN
Product Code. 30311636
Change view
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311636 0.25 g 0.25g
30318661 0.5 g 0.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30311636 Supplier CDN Supplier No. CDND00900.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Acetamide-2,2,2-d3
CAS 23724-60-9
Molecular Formula C2 D3 H2 N O
Quantity 0.25 g
Synonym 2,2,2-Trideuteroacetamide, 2,2,2-Trideuterioacetamide, Acetamide-2,2,2-d3 (7 CI, 8 CI, 9 CI)
InChI Formula InChI=1 S/C2H5NO/c1-2(3)4/h1H3,(H2,3,4)/i1D3
SMILES [2 H]C([2 H])([2 H])C(=O)N
IUPAC Name 2,2,2-trideuterioacetamide
Molecular Weight (g/mol) 62.09
Formula Weight 62.0559 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity