missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropyl-2,2,3,3-d4-amine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Cyclopropyl-2,2,3,3-d4-amine |
| CAS | 1051418-97-3 |
| Molecular Formula | C3 2H4 H3 N |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C3H7N/c4-3-1-2-3/h3H,1-2,4H2/i1D2,2D2 |
| SMILES | [2 H]C1([2 H])C(N)C1([2 H])[2 H] |
| IUPAC Name | 2,2,3,3-tetradeuteriocyclopropan-1-amine |
| Molecular Weight (g/mol) | 61.12 |
| Formula Weight | 61.083 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction