Learn More
Heptanoic-5,5,6,6,7,7,7-d7 Acid, CDN
Product Code. 30318469
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318469
Brand: CDN CDND67780.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Heptanoic Acid-5,5,6,6,7,7,7-D7
CAS 1219802-86-4
Molecular Formula C7 D7 H7 O2
Quantity 0.25 g
Synonym Heptanoic-5,5,6,6,7,7,7-d7 acid, Heptanoic Acid-5,5,6,6,7,7,7-D7, Heptanoic acid-d7, Enanthic acid-d7
InChI Formula InChI=1 S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)/i1D3,2D2,3D2
SMILES [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCCC(=O)O
IUPAC Name 5,5,6,6,7,7,7-heptadeuterioheptanoic acid
Molecular Weight (g/mol) 137.228
Formula Weight 137.1433 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity