missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Tridecyl-d27 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Tridecanol-D27 |
| CAS | 203633-18-5 |
| Molecular Formula | C13 D27 H O |
| Quantity | 0.25 g |
| Synonym | 1-Tridecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,13-d27-ol (9 CI), 1-Tridecanol-D27, 1-Tridecanol-d27, n-Tridecanol-d27, n-Tridecyl-d27 alcohol, Tridecyl alcohol-d27, 1-Hydroxytridecane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,13-d27 |
| InChI Formula | InChI=1 S/C13H28O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14/h14H,2-13H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,13-heptacosadeuteriotridecan-1-ol |
| Molecular Weight (g/mol) | 227.527 |
| Formula Weight | 227.383 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction