Learn More
Quinoxaline-5,6,7,8-d4, CDN
Product Code. 30309019
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30309019
Brand: CDN CDND10330.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Quinoxaline-5,6,7,8-D4
CAS 64252-08-0
Molecular Formula C8 D4 H2 N2
Quantity 0.25 g
Synonym Quinoxaline-5,6,7,8-d4 (9 CI), 1,4-Benzodiazine-5,6,7,8-d4, 1,4-Diazanaphthalene--5,6,7,8-d4, Quinoxaline-d4
InChI Formula InChI=1 S/C8H6N2/c1-2-4-8-7(3-1)9-5-6-10-8/h1-6 H/i1D,2 D,3 D,4 D
SMILES [2 H]c1c([2 H])c([2 H])c2nccnc2c1[2 H]
IUPAC Name 5,6,7,8-tetradeuterioquinoxaline
Molecular Weight (g/mol) 134.1713
Formula Weight 134.0782 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity