missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR beta Antibody (IARC307) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43545
This item is not returnable.
View return policy
Description
TCR beta Monoclonal antibody specifically detects TCR beta in Human samples. It is validated for Flow Cytometry,ELISA,Immunocytochemistry/ Immunofluorescence
Specifications
| TCR beta | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence | |
| T cell receptor beta chain, TCRB, TCRbeta, Tib | |
| Derived from normal B cells in peripheral blood of a Burkitt's lymphoma patient. Screened more than 60 culture supernatants from lymphoblastoid cell lines of normal origin, Epstein-Barr virus (EBV)-carrying Burkitt lymphomas, EBV-negative Burkitt lymphomas and EBV-converted sublines of original EBV-negative Burkitt lymphoma human B-cell lines for their capacity to bind the synthetic peptide autoantigens. One lymphoblastoid B-cell line was identified, IARC307, that produces IgM kappa antibodies which specifically react with the CDR1 peptide corresponding to human Vbeta8.1 gene sequence. (Uniprot# P01850; A0A5B9) | |
| 0.2 mg | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 21577 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| IARC307 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction