missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Di-iso-propylbenzene-d18, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,3-Diisopropylbenzene-D18 |
| CAS | 1398065-59-2 |
| Molecular Formula | C12 D18 |
| Quantity | 0.5 g |
| Synonym | 1,3-Di-iso-propylbenzene-d18, 1,3-Bis[1-(methyl-d3)ethyl-1,2,2,2-d4]benzene-2,4,5,6-d4, 1,3-Diisopropylbenzene-D18, m-Diisopropylbenzene-d18 |
| InChI Formula | InChI=1 S/C12H18/c1-9(2)11-6-5-7-12(8-11)10(3)4/h5-10 H,1-4H3/i1D3,2D3,3D3,4D3,5 D,6 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c(c1[2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,3,5-tetradeuterio-4,6-bis(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)benzene |
| Molecular Weight (g/mol) | 180.3822 |
| Formula Weight | 180.2538 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction