missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromopropane-1,1,1-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Bromopropane-1,1,1-D3 |
| CAS | 688361-45-7 |
| Molecular Formula | C3 D3 H4 Br |
| Quantity | 0.5 g |
| Synonym | Propane-1,1,1-d3, 2-bromo- (9 CI), 2-Bromopropane-1,1,1-d3, 2-Bromo-1,1,1-trideuteriopropane, Isopropyl-d3 bromide, 2-Bromopropane-d3, 2-Bromopropane-1,1,1-D3 |
| InChI Formula | InChI=1 S/C3H7Br/c1-3(2)4/h3H,1-2H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(C)Br |
| IUPAC Name | 2-bromo-1,1,1-trideuteriopropane |
| Molecular Weight (g/mol) | 126.0102 |
| Formula Weight | 124.9919 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction