Learn More
4-Bromofluorobenzene-d4, CDN
Product Code. 30321246
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321246
Brand: CDN CDND55770.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material p-Bromofluorobenzene-d4
CAS 50592-31-9
Molecular Formula C6 D4 Br F
Quantity 0.5 g
Synonym 3-Bromo-6-fluoro-benzene-1,2,4,5-d4, 4-Bromofluorobenzene-d4
InChI Formula InChI=1 S/C6H4BrF/c7-5-1-3-6(8)4-2-5/h1-4 H/i1D,2 D,3 D,4 D
SMILES [2 H]c1c([2 H])c(Br)c([2 H])c([2 H])c1F
IUPAC Name 1-bromo-2,3,5,6-tetradeuterio-4-fluorobenzene
Molecular Weight (g/mol) 179.02
Formula Weight 177.9731 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity