Learn More
Benzophenone-2,3,4,5,6-d5, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | Benzophenone-2,3,4,5,6-d5 |
| CAS | 2694-78-2 |
| Molecular Formula | C13 D5 H5 O |
| Quantity | 0.5 g |
| Synonym | Methanone, phenylphenyl-2,3,4,5,6-d5- (ACI), Benzophenone-2,3,4,5,6-d5 (6 CI, 7 CI, 8 CI), Methanone, phenylphenyl-d5- (9 CI), Phenylphenyl-2,3,4,5,6-d5methanone (ACI), 2,3,4,5,6-Pentadeuteriobenzophenone, 2,3,4,5,6-Pentadeuterobenzophenone, Phenyl(phenyl-d5)methanone, Benzophenone-2,3,4,5,6-D5 |
| InChI Formula | InChI=1 S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10 H/i1D,3 D,4 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)c2ccccc2)c([2 H])c1[2 H] |
| IUPAC Name | (2,3,4,5,6-pentadeuteriophenyl)-phenylmethanone |
| Molecular Weight (g/mol) | 187.249 |
| Formula Weight | 187.105 g/mol |
| Show More |