missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-2,3,4,5,6-d5 Acetate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Benzyl Acetate D5 (phenyl-D5) |
| CAS | 1398065-57-0 |
| Molecular Formula | C9 D5 H5 O2 |
| Quantity | 0.5 g |
| Synonym | Acetic acid benzyl-d5 ester;(Acetoxymethyl)benzene-d5, Benzyl-d5 acetate, Benzyl-2,3,4,5,6-d5 Acetate, Benzyl-d5 ethanoate, Phenyl-d5-methyl acetate, α-Acetoxytoluene-2,3,4,5,6-d5 |
| InChI Formula | InChI=1 S/C9H10O2/c1-8(10)11-7-9-5-3-2-4-6-9/h2-6 H,7H2,1H3/i2D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(COC(=O)C)c([2 H])c1[2 H] |
| IUPAC Name | (2,3,4,5,6-pentadeuteriophenyl)methyl acetate |
| Molecular Weight (g/mol) | 155.2053 |
| Formula Weight | 155.0995 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction