Learn More
Bisphenol A-d16, CDN
Specifications
Specifications
| Chemical Name or Material | Bisphenol A D16 |
| CAS | 96210-87-6 |
| Molecular Formula | C15 2H16 O2 |
| Quantity | 0.5 g |
| Synonym | Phen-2,3,5,6-d4-ol-d, 4,4'-[1-(methyl-d3)ethylidene-2,2,2-d3]bis-, 4,4'-[1-(Methyl-d3)ethylidene-2,2,2-d3]bis[phen-2,3,5,6-d4-ol-d], Bisphenol A-d16, Bisphenol A D16, Perdeuterobisphenol A, 4,4'-Isopropylidenediphenol-d16, 4,4'-Isopropylidenediphenol-d16 |
| InChI Formula | InChI=1 S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17 H,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D/hD2 |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c(c([2 H])c1[2 H])C(c2c([2 H])c([2 H])c(O[2 H])c([2 H])c2[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,4,5-tetradeuterio-3-deuteriooxy-6-[1,1,1,3,3,3-hexadeuterio-2-(2,3,5,6-tetradeuterio-4-deuteriooxyphenyl)propan-2-yl]benzene |
| Molecular Weight (g/mol) | 244.38 |
| Formula Weight | 244.2155 g/mol |
| Show More |