missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Epichlorohydrin-2-d1, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | Epichlorohydrin-2-D |
| CAS | 70735-27-2 |
| Molecular Formula | C3 D H4 Cl O |
| Quantity | 0.5 g |
| Synonym | Oxirane-2-d, 2-(chloromethyl)- (ACI), 2-(Chloromethyl)oxirane-2-d (ACI), Oxirane-d, 2-(chloromethyl)- (9 CI), [2-2 H]-Epichlorohydrin, (±)-Epichlorohydrin-2-d1, Epichlorohydrin-2-D |
| InChI Formula | InChI=1 S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2/i3D |
| SMILES | [2 H]C1(CCl)CO1 |
| IUPAC Name | 2-(chloromethyl)-2-deuteriooxirane |
| Molecular Weight (g/mol) | 93.5304 |
| Formula Weight | 93.0092 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction