Learn More
Pentadecanoic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | Pentadecanoic-2,2-D2 Acid |
| CAS | 64118-45-2 |
| Molecular Formula | C15 D2 H28 O2 |
| Quantity | 0.5 g |
| Synonym | Pentadecanoic-2,2-d2 acid (9 CI), α-Dideuteropentadecanoic acid, Pentadecanoic acid-2,2-d2, 2,2-Dideuteriopentadecanoic acid, Pentadecanoic-2,2-D2 acid |
| InChI Formula | InChI=1 S/C15H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17/h2-14H2,1H3,(H,16,17)/i14D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCCCCCCC)C(=O)O |
| IUPAC Name | 2,2-dideuteriopentadecanoic acid |
| Molecular Weight (g/mol) | 244.41 |
| Formula Weight | 244.237 g/mol |
| Show More |