missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentanedioic-2,2,4,4-d4 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Glutaric-d4 Acid |
| CAS | 19136-99-3 |
| Molecular Formula | C5 D4 H4 O4 |
| Quantity | 0.5 g |
| Synonym | Pentanedioic-2,2,4,4-d4 Acid, Pentanedioic-2,2,4,4-d4 acid (9 CI), Glutaric-2,2,4,4-d4 acid (8 CI), Pentanedioic acid-2,2,4,4-d4, Glutaric acid-2,2,4,4-d4, Glutaric acid-d4, Pentanedioic-d4 acid |
| InChI Formula | InChI=1 S/C5H8O4/c6-4(7)2-1-3-5(8)9/h1-3H2,(H,6,7)(H,8,9)/i2D2,3D2 |
| SMILES | [2 H]C([2 H])(CC([2 H])([2 H])C(=O)O)C(=O)O |
| IUPAC Name | 2,2,4,4-tetradeuteriopentanedioic acid |
| Molecular Weight (g/mol) | 136.14 |
| Formula Weight | 136.0674 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction