Learn More
Propionitrile-3,3,3-d3, CDN
Product Code. 30322130
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322130
Brand: CDN CDND59160.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Propionitrile-3,3,3-D3
CAS 24300-21-8
Molecular Formula C3 D3 H2 N
Quantity 0.5 g
Synonym Propanenitrile-3,3,3-d3 (9 CI), Propionitrile-3,3,3-d3 (8 CI), 3,3,3-Trideuteropropionitrile, Propionitrile-3,3,3-D3
InChI Formula InChI=1 S/C3H5N/c1-2-3-4/h2H2,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])CC#N
IUPAC Name 3,3,3-trideuteriopropanenitrile
Molecular Weight (g/mol) 58.097
Formula Weight 58.061 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity