missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrrolidine-2,2,5,5-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Pyrrolidine-2,2,5,5-D4 |
| CAS | 42403-25-8 |
| Molecular Formula | C4H5D4N |
| Quantity | 0.5 g |
| Synonym | 2,2,5,5-Tetradeuteropyrrolidine |
| InChI Formula | InChI=1 S/C4H9N/c1-2-4-5-3-1/h5H,1-4H2/i3D2,4D2 |
| SMILES | [2 H]C1([2 H])CCC([2 H])([2 H])N1 |
| IUPAC Name | pyrrolidine-2,2,5,5-d4 |
| Molecular Weight (g/mol) | 75.15 |
| Formula Weight | 75.1 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction