missing translation for 'onlineSavingsMsg'
Learn More

Chlorothiazide-13C,15N2, TRC

Product Code. 30011624
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011624 0.5 mg 0.5mg
30133221 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011624 Supplier Toronto Research Chemicals Supplier No. TRCC3800020.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189440-79-6
Molecular Formula C613CH6ClN15N2O4S2
Molecular Weight (g/mol) 298.7
Synonym 6-Chloro-7-sulfamyl-1,2,4-benzothiadiazine-1,1-dioxide-13C,15N2, 6-Chloro-2H-1,2,4-benzothiadiazine-7-sulfonamide-13C,15N2 1,1-dioxide, Chlorthiazide-13C,15N2;Chlotride-13C,15N2;Diuril-13C,15N2, Saluric-13C,15N2;
IUPAC Name 6-chloro-4H-benzo[e][1,2,4]thiadiazine-7-sulfonamide-3-13C-2-15N-15N 1,1-dioxide
SMILES ClC1=C(S([15NH2])(=O)=O)C=C2C(N[13CH]=[15N]S2(=O)=O)=C1

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C7H6ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-3H,(H,10,11)(H2,9,12,13)/i3+1,9+1,11+1
Formula Weight 297.95
Percent Purity >95%
Chemical Name or Material Chlorothiazide-13C,15N2
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.