Learn More
Valganciclovir N3,N3'-Methylene Dimer (>90%), TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | Valganciclovir N3,N3'-Methylene Dimer |
| CAS | 1401661-96-8 |
| Molecular Formula | C29 H44 N12 O10 |
| Quantity | 0.5 mg |
| Synonym | L-Valine','1,1'-[methylenebis[imino(1,6-dihydro-6-oxo-9H-purine-2,9-diyl)methyleneoxy[2-(hydroxymethyl)-2,1-ethanediyl]]] ester, [2-[[2-[[[9-[[1-[[(2S)-2-amino-3-methyl-butanoyl]oxymethyl]-2-hydroxy-ethoxy]methyl]-6-oxo-3H-purin-2-yl]amino]methylamino]-6-oxo-3H-purin-9-yl]methoxy]-3-hydroxy-propyl] (2S)-2-amino-3-methyl-butanoate, 1,1'-[Methylenebis[imino(1,6-dihydro-6-oxo-9H-purine-2,9-diyl)methyleneoxy[2-(hydroxymethyl)-2,1-ethanediyl]]] Ester L-Valine |
| InChI Formula | InChI=1S/C29H44N12O10/c1-14(2)18(30)26(46)48-7-16(5-42)50-12-40-10-34-20-22(40)36-28(38-24(20)44)32-9-33-29-37-23-21(25(45)39-29)35-11-41(23)13-51-17(6-43)8-49-27(47)19(31)15(3)4/h10-11,14-19,42-43H,5-9,12-13,30-31H2,1-4H3,(H2,32,36,38,44)(H2,33,37,39,45)/t16?,17?,18-,19-/m0/s1 |
| SMILES | CC(C)[C@H](N)C(=O)OCC(CO)OCn1cnc2C(=O)N=C(NCNC3=NC(=O)c4ncn(COC(CO)COC(=O)[C@@H](N)C(C)C)c4N3)Nc12 |
| IUPAC Name | [2-[[2-[[[9-[[1-[(2S)-2-amino-3-methylbutanoyl]oxy-3-hydroxypropan-2-yl]oxymethyl]-6-oxo-3H-purin-2-yl]amino]methylamino]-6-oxo-3H-purin-9-yl]methoxy]-3-hydroxypropyl] (2S)-2-amino-3-methylbutanoate |
| Molecular Weight (g/mol) | 720.7341 |
| Formula Weight | 720.3303 |