missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Trimethyl-1H-benzo[e]indole, 97%

Product Code. 11404827
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404827 1 g 1g
11414827 5 g 5g
11424827 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404827 Supplier Thermo Scientific Alfa Aesar Supplier No. B21804.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 41532-84-7 | C15H15N | 209.29 g/mol

Reactant used for preparation of fluorescent probes for imaging of tumors, hypoxia-sensitive fluorescent probes for real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of fluorescent probes for imaging of tumors in vivo, hypoxia-sensitive fluorescent probes for in vivo real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41532-84-7
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.29
MDL Number MFCD00082627
InChI Key WJZSZXCWMATYFX-UHFFFAOYSA-N
Synonym 1,1,2-trimethyl-1h-benzo e indole, 1,1,2-trimethyl-1h-benz e indole, 1,1,2-trimethylbenz e indole, 2,3,3-trimethyl-4,5-benzo-3h-indole, 2,3,3-trimethylbenzoindolenine, 1,1,2-trimethylbenzo e indole, 1h-benz e indole, 1,1,2-trimethyl, 2,3,3-trimethyl-1h-benz e indolenine, 1,1,2-trimethylhydrobenzo e indole
PubChem CID 170530
IUPAC Name 1,1,2-trimethylbenzo[e]indole
SMILES CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C

Specifications

Melting Point 114°C to 119°C
Quantity 1 g
Beilstein 153709
Solubility Information Slightly soluble in water.
Formula Weight 209.29
Percent Purity 97%
Chemical Name or Material 1,1,2-Trimethyl-1H-benzo[e]indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.