Learn More
1,1,3-Trichloro-1-propene, TRC
Specifications
Specifications
| Chemical Name or Material | 1,1,3-Trichloro-1-propene |
| CAS | 2567-14-8 |
| Molecular Formula | C3H3Cl3 |
| Quantity | 10 mg |
| Synonym | 1,1,3-Trichloropropene, 1,3,3-Trichloro-2-propene, 3,3-Dichloroallyl Chloride, HCC 1240za, 1,1,3-Trichloroprop-1-ene |
| InChI Formula | InChI=1S/C3H3Cl3/c4-2-1-3(5)6/h1H,2H2 |
| SMILES | ClC/C=C(Cl)/Cl |
| IUPAC Name | 1,1,3-trichloroprop-1-ene |
| Molecular Weight (g/mol) | 145.41 |
| Formula Weight | 143.93 |