missing translation for 'onlineSavingsMsg'
Learn More

1,1'-[(5E)-5-Methyl-7-oxo-5-undecene-1,11-diyl] Bis[Theobromine], TRC

Product Code. 30475317
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475317 10 mg 10mg
30474996 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30475317 Supplier Toronto Research Chemicals Supplier No. TRCM27000010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1'-[(5E)-5-Methyl-7-oxo-5-undecene-1,11-diyl] Bis[Theobromine]
CAS 874747-30-5
Purity Grade Notes HPLC
Molecular Formula C26 H34 N8 O5
Quantity 10 mg
Synonym 3,7-Dihydro-3,7-dimethyl-1-[(5E)-5-methyl-7-oxo-11-(2,3,6,7-tetrahydro-3,7-dimethyl-2,6-dioxo-1H-purin-1-yl)-5-undecen-1-yl]-1H-purine-2,6-dione, 1,1'-[(5E)-5-Methyl-7-oxo-5-undecene-1,11-diyl]bis[3,7-dihydro-3,7-dimethyl-1H-purine-2,6-dione, Pentoxifylline EP Impurity J
InChI Formula InChI=1S/C26H34N8O5/c1-17(10-6-8-12-33-23(36)19-21(27-15-29(19)2)31(4)25(33)38)14-18(35)11-7-9-13-34-24(37)20-22(28-16-30(20)3)32(5)26(34)39/h14-16H,6-13H2,1-5H3/b17-14+
SMILES CN1C(=O)N(CCCC\C(=C\C(=O)CCCCN2C(=O)N(C)c3ncn(C)c3C2=O)\C)C(=O)c4c1ncn4C
IUPAC Name 1-[(E)-11-(3,7-dimethyl-2,6-dioxopurin-1-yl)-5-methyl-7-oxoundec-5-enyl]-3,7-dimethylpurine-2,6-dione
Molecular Weight (g/mol) 538.6
Formula Weight 538.2652
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.