Learn More
[1,1'-Bicyclohexyl]-1-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (1,1'-Bicyclohexyl)-1-carboxylic Acid |
| CAS | 60263-54-9 |
| Molecular Formula | C13 H22 O2 |
| Quantity | 1 g |
| Synonym | [1,1'-Bicyclohexyl]-1-carboxylic acid, [Bicyclohexyl]-1-carboxylic acid (6CI,7CI), Bicyclohexyl-1-carboxylic acid, Dicycloverine EP impurity A |
| InChI Formula | InChI=1S/C13H22O2/c14-12(15)13(9-5-2-6-10-13)11-7-3-1-4-8-11/h11H,1-10H2,(H,14,15) |
| SMILES | OC(=O)C1(CCCCC1)C2CCCCC2 |
| IUPAC Name | 1-cyclohexylcyclohexane-1-carboxylic acid |
| Molecular Weight (g/mol) | 210.31 |
| Formula Weight | 210.162 |